Beschreibung
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility (14170-120). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility (14170-120). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Spezifikation
Spezifikation
| Certifications/Compliance | ISO 13485 standards, FDA |
| Content And Storage | 15 to 30°C |
| Description | Hanks' Balanced Salt Solution, 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Glucose, Sodium Bicarbonate, Phenol Red, 36 Months Shelf Life, For Mammalian Cell Culture |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Hanks' Balanced Salt Solution |
| With Additives | Glucose, Sodium Bicarbonate, Phenol Red |
| Without Additives | No Calcium, No Magnesium |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 7.0 to 7.4 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?