Learn More
Gibco™ HBSS, with calcium and magnesium; no phenol red
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HBSS product made in our Grand Island facility (14025-134). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HBSS product made in our Grand Island facility (14025-134). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
Specifications
| Certifications/Compliance | ISO 13485 standards, FDA |
| Content And Storage | 15 to 30°C |
| Description | Hanks' Balanced Salt Solution, 10 x 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Calcium, Magnesium, Glucose, Sodium Bicarbonate, 36 Months Shelf Life, For Mammalian Cell Culture |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Hanks' Balanced Salt Solution |
| With Additives | Calcium, Magnesium, Glucose, Sodium Bicarbonate |
| Without Additives | No Phenol Red |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 7.0 to 7.4 |
| Show More |