Learn More
Gibco™ HBSS, with calcium and magnesium; no phenol red
GREENER_CHOICE
Product Code. 11550456
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
Product Code. 11550456
Brand: Gibco™ 14025050
HBSS, with calcium and magnesium. No phenol red.
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HBSS product made in our Grand Island facility (14025-134). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 13485 standards, FDA
Content And Storage Store at 15–30°C
Description Hanks' Balanced Salt Solution, 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Calcium, Magnesium, Glucose, Sodium Bicarbonate, 36 Months Shelf Life, For Mammalian Cell Culture
Concentration 1X
Form Liquid
Product Type Hanks' Balanced Salt Solution
With Additives Calcium, Magnesium, Glucose, Sodium Bicarbonate
Without Additives No Phenol Red
For Use With (Application) Mammalian Cell Culture
pH 6.7 to 7.8
Quantity 500 mL
Regulatory Status For in-vitro diagnostic use