missing translation for 'onlineSavingsMsg'
Learn More

Hashishene, TRC

Product Code. 17959334
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959334 100 mg 100mg
17979434 1 g 1g
17919444 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17959334 Supplier Toronto Research Chemicals Supplier No. TRCH105025100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16626-39-4
Molecular Formula C10 H16
Molecular Weight (g/mol) 136.234
Synonym Bicyclo[2.1.1]hexane, 5,5-dimethyl-1-vinyl- (7CI,8CI), 1-Ethenyl-5,5-dimethylbicyclo[2.1.1]hexane, 1-Vinyl-5,5-dimethyl[2.1.1]bicyclohexane, Hashishene, Bicyclo[2.1.1]hexane, 1-ethenyl-5,5-dimethyl-
IUPAC Name 4-ethenyl-5,5-dimethylbicyclo[2.1.1]hexane
SMILES CC1(C)C2CCC1(C2)C=C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H16/c1-4-10-6-5-8(7-10)9(10,2)3/h4,8H,1,5-7H2,2-3H3
Formula Weight 136.125
Chemical Name or Material Hashishene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.