CAS RN 16626-39-4
CAS RN 16626-39-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Hashishene, TRC
CAS: 16626-39-4 Molecular Formula: C10 H16 Molecular Weight (g/mol): 136.234 Synonym: Bicyclo[2.1.1]hexane, 5,5-dimethyl-1-vinyl- (7CI,8CI),1-Ethenyl-5,5-dimethylbicyclo[2.1.1]hexane,1-Vinyl-5,5-dimethyl[2.1.1]bicyclohexane,Hashishene,Bicyclo[2.1.1]hexane, 1-ethenyl-5,5-dimethyl- IUPAC Name: 4-ethenyl-5,5-dimethylbicyclo[2.1.1]hexane SMILES: CC1(C)C2CCC1(C2)C=C
2
Hashishene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Hashishene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.