Learn More
Hanna Instruments™ Hardness Reagent Water Testing Kit
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Description
- Pre-made reagents for ease of use
- All reagents marked with expiration date and lot number for traceability
- Manual titration performed with calmagite indicator
Specifications
Specifications
| No. of Tests | 100 |
| Quantity | 1 Each |
| Range | 0 to 300mg/L |
| Test Method | Titration |
| Type | Hardness Test Kit |