Learn More
Sampling Systems™ Hand Scraper
Product Code. 17962062 Shop All Sampling Systems Products
Change view
Click to view available options
Color:
Blue
Red
White
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17962062 Red Each
17952062 White Each
17972062 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17962062 Supplier Sampling Systems™ Supplier No. 8336R1
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 02-12-2026
These robust and sturdy hand scrapers are ideal for easily scraping large areas due to their huge blade size and handle. The performance is further enhanced by the fact that the scrapers are made out of polypropylene for very good chemical resistance.
  • Robust and incorporates a large, sturdy handle
  • Large 250 mm wide blade
  • Comes in 3 distinctive colors
Specifications
Product Type Scraper
Color-coded No
Handle Material Polypropylene
Handle Type Polypropylene
Tip Type Flat
Scent Regular scent
Sterility Non-sterile
Autoclavable Autoclavable
Color Red
Disposable Reusable
Material Polypropylene
Size 250 mm