Learn More
Tork Hand Cleaning Wet Wipe Refill W14
GREENER_CHOICE
Product Code. 30243701 Shop All Tork Products
Change view
Click to view available options
Color:
White
Unit Size:
4 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243701 White 4 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30243701 Supplier Tork Supplier No. 190692
  • ONLINE EXCLUSIVE PRICE
    124.00 EUR
    120.83 EUR / 4 rolls
    Save 3.17 EUR
    2% Off
View all promotions
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
The Tork Hand Cleaning Wet Wipes are convenient and efficient for hand cleaning when water is not available. The product makes refilling easy and efficient.
  • Remove oil, grease, lubricants and dirt with ease, thanks to the strong texture and high absorbency of these wet wipes.
  • Clean hands effectively with these professional-quality wipes.
  • Avoid potential irritations with wipes that are free of fragrance and kind to your skin.
  • Refilling is easy and efficient with our bucket-style dispenser.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wet Wipe
Length (English) 17.17 yd.
Length (Metric) 15.7 m
Width (English) 5.31 in.
Width (Metric) 13.5 cm
Certifications/Compliance FSC MIX Credit
Color White
Packaging Quantity 4 x 58 Wipes
Ply 1-ply
Certifications/Compliance (Europe) FSC MIX Credit