Learn More
Hamilton™ PB600-1 Repeating Dispenser
Product Code. 10707971
Change view
Click to view available options
For Use With (Equipment):
Microliter and Gastight Syringes (25μL to 2.5 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10707971 Microliter and Gastight Syringes (25μL to 2.5 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10707971 Supplier Hamilton™ Supplier No. 83700
  • ONLINE EXCLUSIVE PRICE
    229.00 EUR
    179.46 EUR / Pack of 1
    Save 49.54 EUR
    21% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Repeating dispenser for Hamilton Company syringes
  • Designed to consistently dispense a sample up to 50 times with the push of a button
  • Dispensers work with Hamilton Company Microliter and Gastight syringes in a 25μL to 2.5mL volume
Specifications
Type Repeating Dispenser
For Use With (Equipment) Microliter and Gastight Syringes (25μL to 2.5 mL)