Learn More
Hamilton™ Tungsten Cleaning Wires
Product Code. 10642003
Change view
Click to view available options
For Use With (Equipment):
22, 23G and Larger Needle, Syringe and Needle Cleaning Kit
22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit
23s Needle, Syringe and Needle Cleaning Kit
24 to 26G Needle, Syringe and Needle Cleaning Kit
26s and 31 to 33G Needle, Syringe and Needle Cleaning Kit
27G Needle, Syringe and Needle Cleaning Kit
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10642003 22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit Pack of 10
10464302 23s Needle, Syringe and Needle Cleaning Kit Each
11588853 24 to 26G Needle, Syringe and Needle Cleaning Kit Pack of 10
10612503 26s and 31 to 33G Needle, Syringe and Needle Cleaning Kit Pack of 10
10345382 27G Needle, Syringe and Needle Cleaning Kit Pack of 10
10427171 22, 23G and Larger Needle, Syringe and Needle Cleaning Kit Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10642003 Supplier Hamilton™ Supplier No. 18301
  • ONLINE EXCLUSIVE PRICE
    26.48 EUR
    23.79 EUR / Pack of 10
    Save 2.69 EUR
    10% Off
Estimated Shipment: 08-12-2026
For mechanical cleaning to remove needle obstructions.
  • Can be threaded through the needle to clear blockages
  • For a variety of needle gauges
  • Provided in packs of 10
Order Info
Ten wires per pack.
Specifications
Material Tungsten
Diameter (Metric) Outer 0.126 mm
Length (English) 7 in.
Length (Metric) 17.78 cm
Product Type Cleaning Wire
Packaging 10/Pk.
For Use With (Equipment) 22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit