Learn More
Hamilton™ Micro Pipet Disposable Tips
Product Code. 11512984
Change view
Click to view available options
For Use With (Equipment):
Microsyringe Pipet Model 701
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11512984 Microsyringe Pipet Model 701 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11512984 Supplier Hamilton™ Supplier No. 84255
Estimated Shipment: 18-01-2027
Microsyringe Pipets have special 3/4 in. electrotapered needle for easy attachment of PTFE tips
  • Disposable PTFE tips that are to be used with the Microsyringe Pipet (13-684-112A)
  • Sample only wets disposable PTFE tips, eliminating sample cross-contamination
  • Spring-loaded plunger and adjustable stops ensure excellent reproducibility
  • Adjustable stop allows transfer of samples from .2 to 3μL
Specifications
Material PTFE
Disposable Single-use
Volume (Metric) 0.2 to 3 μL
For Use With (Equipment) Microsyringe Pipet Model 701