Learn More
MedChemExpress H2DCFDA
Specifications
Specifications
| Content And Storage | -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 125 mg/mL (256.52 mM; Need ultrasonic) ∣ Ethanol : 20 mg/mL (41.04 mM; Need ultrasonic) |
| Target | HBV |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 487.29 g/mol |
| CAS | 4091-99-0 |
| Synonym | DCFH-DA ∣ 2',7'-Dichlorodihydrofluorescein diacetate |