missing translation for 'onlineSavingsMsg'
Learn More

H-HoArg-OH, MedChemExpress

Product Code. 16439336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439336 Supplier Medchem Express Supplier No. HYW008385

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress H-HoArg-OH, a homologue arginine, is a strong inhibitor of human bone and liver alkaline phosphatase.

H-HoArg-OH, a homologue arginine, is a strong inhibitor of human bone and liver alkaline phosphatase.

Specifications

Chemical Name or Material H-HoArg-OH
CAS 156-86-5
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C7H16N4O2
Solubility Information H2O : 100 mg/mL (531.26 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES [H][C@](N)(CCCCNC(N)=N)C(O)=O
Molecular Weight (g/mol) 188.23
Quantity 100 mg
Formula Weight 188.23
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.