Learn More
SP Bel-Art™ H-B DURAC Liquid Crystal Body Forehead Thermometer 100/Pk.
Product Code. 16626652
Change view
Click to view available options
Length (Metric):
45 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
16626652 45 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16626652 Supplier SP Bel-Art™ Supplier No. B620100100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Non-invasive body temperature measurement is ideal for workforce personnel.
  • Color-coded readings allow for quick evaluation of human temperature
  • Easy to use - simply peel off backing and stick to forehead or hand
  • Can be self-applied to maintain social distancing during temperature testing
  • Latex-free
  • Utilizes Thermocratic Liquid Crystal technology
  • Disposable
  • Instructions included
Specifications
Disposable Single-use
Length (Metric) 45 mm
Max. Temperature (Metric) 40°C
Min. Temperature (Metric) 34°C