Learn More
Thermo Scientific™ H-12000 Bioprocessing Swinging-Bucket Rotor
Product Code. 10308369 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
6 x 2400 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10308369 6 x 2400 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10308369 Supplier Thermo Scientific™ Supplier No. 77080
Integrate the Thermo Scientific™ H-12000 Bioprocessing Swinging-Bucket Rotor with Thermo Scientific™ Sorvall™ RC12BP low-speed centrifuges.
TRUSTED_SUSTAINABILITY
Specifications
Description H-12000 Bioprocessing Swinging-Bucket Rotor
Material Stainless Steel Rotor Body, Aluminum Bucket
Type Swinging Bucket Rotor
Capacity 6 x 2400 mL
Max. RCF 7,340 x g
Max. Speed 4,700 rpm
Radius (Metric) 297.2 mm
Radius (Metric) Min. 141 mm
K Factor 8117
For Use With (Equipment) Thermo Scientific Sorvall RC 12 Series Large Capacity Centrifuges
Angle 90°
Min. RCF 3,610 x g