missing translation for 'onlineSavingsMsg'
Learn More

GW311616, MedChemExpress

Product Code. p-200101605
Encompass
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496976 100 mg 100mg
16459437 5 mg 5mg
16422967 50 mg 50mg
16416908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496976 Supplier Medchem Express Supplier No. HY15891

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress GW-311616 is a potent, orally bioavailable, long duration and selective human neutrophil elastase (HNE) inhibitor with IC50 value of 22 nM and Ki value of 0.31 nM.

GW-311616 is a potent, orally bioavailable, long duration and selective human neutrophil elastase (HNE) inhibitor with IC50 value of 22 nM and Ki value of 0.31 nM.

Specifications

Chemical Name or Material GW311616
CAS 198062-54-3
Color Brown
Purity Grade Notes Research
Molecular Formula C19H31N3O4S
Quantity 100 mg
Solubility Information DMSO : 66.67 mg/mL (167.71 mM; Need ultrasonic)
SMILES O=C1[C@@H](C(C)C)[C@@](N(C(/C=C/CN2CCCCC2)=O)CC3)([H])[C@]3([H])N1S(=O)(C)=O
Molecular Weight (g/mol) 397.53
Formula Weight 397.53
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.