Learn More
Kartell™ Set of 7 Adapters (Guko)
Product Code. 10737862
Change view
Click to view available options
Type:
Adapters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10737862 Adapters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10737862 Supplier Kartell™ Supplier No. 0042000
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Kartell™ Set of 7 Adapters (Guko) offers support to a Buchner Funnel and includes adapters with a variety of dimensions.
  • Supports for Buchner Funnels
  • Constructed of neoprene rubber.
  • Dimensions for adaptor 1:
    • Exterior dia. 21 x 11mm, Interior dia. 17 x 7mm, Height 21mm
  • Dimensions for adaptor 2:
    • Exterior dia. 27 x 16mm, Interior dia. 22 x 11mm, Height 22mm
  • Dimensions for adaptor 3:
    • Exterior dia. 37 x 22mm, Interior dia. 31 x 16mm, Height 25mm
  • Dimensions for adaptor 4:
    • Exterior dia. 46 x 29mm, Interior dia. 39 x 22mm, Height 29mm
  • Dimensions for adaptor 5:
    • Exterior dia. 58 x 38mm, Interior dia. 50 x 30mm, Height 35mm
  • Dimensions for adaptor 6:
    • Exterior dia. 69 x 45mm, Interior dia. 60 x 36mm, Height 40mm
  • Dimensions for adaptor 7:
    • Exterior dia. 86 x 57mm, Interior dia. 75 x 46mm, Height 45mm
Specifications
Color Gray
For Use With (Equipment) Buchner Funnel
Quantity 7 Adapters
Material Neoprene
Type Adapters