missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-monophosphate, Free acid, ≥98%, MP Biomedicals™

Product Code. 11394359 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394359 100 mg 100mg
11304369 500 mg 500mg
11314369 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394359 Supplier MP Biomedicals Supplier No. 0210190880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85-32-5
Molecular Formula C10H14N5O8P
Molecular Weight (g/mol) 363.223
InChI Key RQFCJASXJCIDSX-UUOKFMHZSA-N
Synonym 5'-guanylic acid, guanosine monophosphate, guanylic acid, guanosine-5'-monophosphate, guanosine 5'-monophosphate, 5'-gmp, guanosine 5'-phosphate, guanylate, guanidine monophosphate, guanosine-phosphate
PubChem CID 6804
ChEBI CHEBI:17345
IUPAC Name [(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)NC(=NC2=O)N

Specifications

Color White
Assay Percent Range ≥98%
Quantity 100 mg
Formula Weight 363.2
Physical Form Powder
Chemical Name or Material Guanosine-5'-monophosphate, FTIR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.