Learn More
Guanosine-5'-monophosphate disodium salt hexahydrate, ≥98%, MP Biomedicals™
Product Code. 11477790 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477790 5 g 5g
11467790 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11477790 Supplier MP Biomedicals Supplier No. 0210070205
  • ONLINE EXCLUSIVE PRICE
    150.00 EUR
    142.80 EUR / 5g
    Save 7.20 EUR
    4% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Guanosine 5'-monophosphate (GMP) is a ribonucleoside monophospate which upon phosphorylation to GTP becomes incorporated into ribonucleic acids (RNAs) by various RNA polymerase(s)
Chemical Identifiers
CAS 5550-12-9
Molecular Formula C10H12N5Na2O8P
Molecular Weight (g/mol) 407.19
MDL Number MFCD00151238
InChI Key PVBRXXAAPNGWGE-KHRSEZDTNA-L
Synonym guanosine 5'-monophosphate disodium salt, 5'-guanylic acid disodium salt, disodium guanylate, disodium 5'-guanylate, sodium guanylate, disodium gmp, disodium 5'-gmp, gmp disodium salt, 5'-guanylic acid, disodium salt, 5'-gmp disodium salt
PubChem CID 21712
IUPAC Name disodium [(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1
Specifications
Color White
Assay Percent Range ≥98%
Quantity 5 g
Formula Weight 407.2 (Anhydrous)
Physical Form Powder
Chemical Name or Material Guanosine-5'-monophosphate disodium salt hexahydrate