missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-monophosphate disodium salt hexahydrate, ≥98%, MP Biomedicals™

Product Code. 11467790 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467790 1 g 1g
11477790 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467790 Supplier MP Biomedicals Supplier No. 0210070201
  • ONLINE EXCLUSIVE PRICE
    55.15 EUR
    52.50 EUR / 1g
    Save 2.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Guanosine 5'-monophosphate (GMP) is a ribonucleoside monophospate which upon phosphorylation to GTP becomes incorporated into ribonucleic acids (RNAs) by various RNA polymerase(s)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5550-12-9
Molecular Formula C10H12N5Na2O8P
Molecular Weight (g/mol) 407.19
MDL Number MFCD00151238
InChI Key PVBRXXAAPNGWGE-KHRSEZDTNA-L
Synonym guanosine 5'-monophosphate disodium salt, 5'-guanylic acid disodium salt, disodium guanylate, disodium 5'-guanylate, sodium guanylate, disodium gmp, disodium 5'-gmp, gmp disodium salt, 5'-guanylic acid, disodium salt, 5'-gmp disodium salt
PubChem CID 21712
IUPAC Name disodium [(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Assay Percent Range ≥98%
Quantity 1 g
Formula Weight 407.2 (anhydrous)
Physical Form Powder
Chemical Name or Material Guanosine-5'-monophosphate disodium salt hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.