missing translation for 'onlineSavingsMsg'
Learn More

Guaiazulene, 98+%

Product Code. p-7023792
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493268 5 g 5g
11403278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493268 Supplier Thermo Scientific Alfa Aesar Supplier No. A17690.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 489-84-9 | C15H18 | 198.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is employed as antimicrobial agent and perfuming agent in cosmetics industry.

Solubility
Soluble in alcohol, water, 0.1115 mg/L @ 25°C (est).

Notes
Store in dark. Protect from heat, oxidizing agents and heat. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 489-84-9
Molecular Formula C15H18
Molecular Weight (g/mol) 198.309
MDL Number MFCD00003811
InChI Key FWKQNCXZGNBPFD-UHFFFAOYSA-N
Synonym guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen
PubChem CID 3515
ChEBI CHEBI:5550
IUPAC Name 1,4-dimethyl-7-propan-2-ylazulene
SMILES CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C

Specifications

Melting Point 31°C to 33°C
Density 0.976
Boiling Point 152°C to 154°C (7 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1365001
Sensitivity Light sensitive
Merck Index 14,4554
Solubility Information Soluble in alcohol,water,0.1115mg/L @ 25°C (est).
Formula Weight 198.31
Percent Purity ≥98%
Chemical Name or Material Guaiazulene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.