missing translation for 'onlineSavingsMsg'
Learn More

GTx-007, MedChemExpress

Product Code. 16444928
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444928 10 mg 10mg
16435706 100 mg 100mg
16427677 5 mg 5mg
16421177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16444928 Supplier Medchem Express Supplier No. HY12023
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GTx-007 (S-4) is an orally active and selective nonsteroidal androgen receptor (AR) modulator (SARM) and a partial agonist, with Ki of 4 nM. GTx-007 (S-4) is identified as SARMs with potent and tissue-selective in vivo pharmacological activity.

GTx-007 (S-4) is an orally active and selective nonsteroidal androgen receptor (AR) modulator (SARM) and a partial agonist, with Ki of 4 nM. GTx-007 (S-4) is identified as SARMs with potent and tissue-selective in vivo pharmacological activity.

Specifications

Chemical Name or Material GTx-007
CAS 401900-40-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H18F3N3O6
Quantity 10 mg
Synonym S-4
Solubility Information DMSO : ≥ 100 mg/mL (226.57 mM)
SMILES O=C([C@@](O)(COC1=CC=C(C=C1)NC(C)=O)C)NC2=CC=C(C(C(F)(F)F)=C2)[N+]([O-])=O
Molecular Weight (g/mol) 441.36
Formula Weight 441.36
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.