Learn More
GSK369796 Dihydrochloride, MedChemExpress
Product Code. 16490146
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490146 2 mg 2mg
16484978 10 mg 10mg
16475746 100 mg 100mg
16451227 50 mg 50mg
16407717 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16490146 Supplier Medchem Express Supplier No. HY12082A
 Restricted Item
Estimated Shipment: 10-11-2026
MedChemExpress GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM.
GSK369796 Dihydrochloride is an affordable and effective antimalarial and inhibits hERG potassium ion channel repolarization with an IC50 of 7.5 μM.
Specifications
Chemical Name or Material GSK369796 Dihydrochloride
Physical Form Solid
CAS 1010411-21-8
Color Yellow
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C20H24Cl3N3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (233.22 mM; Need ultrasonic) ∣H2O : 50 mg/mL (116.61 mM; Need ultrasonic)
SMILES OC1=CC(NC2=CC=NC3=CC(Cl)=CC=C23)=CC=C1CNC(C)(C)C.[H]Cl.[H]Cl
Molecular Weight (g/mol) 428.78
Formula Weight 428.78
Percent Purity 98.32%
Grade Research