Learn More
GSK256066, MedChemExpress
Product Code. 16454996
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454996 100 mg 100mg
16450347 50 mg 50mg
16426707 5 mg 5mg
16403998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16454996 Supplier Medchem Express Supplier No. HY10469
MedChemExpress GSK256066 is a selective and high-affinity phosphodiesterase 4 (PDE4) inhibitor, with an IC50 of 3.2 pM for PDE4B. GSK256066 is developed for the research of chronic obstructive pulmonary disease.
GSK256066 is a selective and high-affinity phosphodiesterase 4 (PDE4) inhibitor, with an IC50 of 3.2 pM for PDE4B. GSK256066 is developed for the research of chronic obstructive pulmonary disease.
Specifications
Chemical Name or Material GSK256066
CAS 801312-28-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H26N4O5S
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (48.21 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N)C1=C(C2=CC(S(=O)(C3=CC=CC(C(N(C)C)=O)=C3)=O)=CC(C)=C2N=C1)NC4=CC=CC(OC)=C4
Molecular Weight (g/mol) 518.58
Formula Weight 518.58
Percent Purity 98.83%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation