Learn More
GSK2018682, MedChemExpress
Product Code. 16480248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480248 5 mg 5mg
16457626 100 mg 100mg
16443646 25 mg 25mg
16437788 10 mg 10mg
16423747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16480248 Supplier Medchem Express Supplier No. HY19511
 Restricted Item
Estimated Shipment: 08-10-2026
MedChemExpress GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis.
GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis.
Specifications
Chemical Name or Material GSK2018682
CAS 1034688-30-6
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClN4O4
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (283.52 mM)
SMILES O=C(O)CCCN1C=CC2=C1C=CC=C2C3=NOC(C4=CC(Cl)=C(OC(C)C)N=C4)=N3
Molecular Weight (g/mol) 440.88
Formula Weight 440.88
Percent Purity 98.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation