Learn More
GSK0660, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms.
Specifications
Specifications
| Chemical Name or Material | GSK0660 |
| CAS | 1014691-61-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C19H18N2O5S2 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 49 mg/mL (117.09 mM) |
| SMILES | O=C(C1=C(S(=O)(NC2=CC=C(NC3=CC=CC=C3)C=C2OC)=O)C=CS1)OC |
| Molecular Weight (g/mol) | 418.49 |
| Formula Weight | 418.49 |
| Show More |