missing translation for 'onlineSavingsMsg'
Learn More

GSK-J1 lithium salt, MedChemExpress

Product Code. p-200101917
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482797 50 mg 50mg
16466718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482797 Supplier Medchem Express Supplier No. HY15648D
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK-J1 lithium salt is a potent inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A, with IC50 of 60 nM towards KDM6B.

GSK-J1 lithium salt is a potent inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A, with IC50 of 60 nM towards KDM6B.

Specifications

Chemical Name or Material GSK-J1 lithium salt
Physical Form Solid
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H22LiN5O2
For Use With (Application) Neuroscience-Neuromodulation
SMILES [Li]OC(CCNC1=NC(C2=NC=CC=C2)=NC(N(CC3)CCC4=C3C=CC=C4)=C1)=O
Molecular Weight (g/mol) 395.38
Formula Weight 395.38
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.