missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-J1 lithium salt, MedChemExpress
Description
GSK-J1 lithium salt is a potent inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A, with IC50 of 60 nM towards KDM6B.
Specifications
Specifications
| Chemical Name or Material | GSK-J1 lithium salt |
| Physical Form | Solid |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H22LiN5O2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| SMILES | [Li]OC(CCNC1=NC(C2=NC=CC=C2)=NC(N(CC3)CCC4=C3C=CC=C4)=C1)=O |
| Molecular Weight (g/mol) | 395.38 |
| Formula Weight | 395.38 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?