missing translation for 'onlineSavingsMsg'
Learn More

GSK 525768A, MedChemExpress

Product Code. p-200101003
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486076 100 mg 100mg
16475498 10 mg 10mg
16431677 50 mg 50mg
16418217 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486076 Supplier Medchem Express Supplier No. HY13032A
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK 525768A is the inactive enantiomer of GSK525762A. GSK 525768A has no activity towards BET.

GSK 525768A is the inactive enantiomer of GSK525762A. GSK 525768A has no activity towards BET.

Specifications

Chemical Name or Material GSK 525768A
CAS 1260530-25-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H22ClN5O2
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (235.90 mM)
SMILES O=C(NCC)C[C@@H]1C2=NN=C(C)N2C3=CC=C(OC)C=C3C(C4=CC=C(Cl)C=C4)=N1
Molecular Weight (g/mol) 423.9
Formula Weight 423.9
Percent Purity 96.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.