missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 143

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GSK 143 is a spleen tyrosine kinase (Syk) inhibitor (pIC50 = 7.5). Also inhibits phosphorylated Erk (pIC50 = 7.1). Decreases cell number and viability in chronic lymphocytic leukemia (CCL) cell lines.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C17H22N6O2.2HCl |
| Formula Weight | 415.32 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction