Learn More
AL Glove Systems GSB Isolator Box Gloves
Product Code. 31337508
Change view
Click to view available options
Size:
Large, 9.5
Medium, 8
X-Large, 11
Length (Metric):
800 mm
920 mm
Thickness (Metric):
0.4 mm
0.6 mm
Diameter (Metric) Cuff:
160 mm
180 mm
200 mm
225 mm
250 mm
300 mm
34 product options available for selection
Product selection table with 34 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Length (Metric) Thickness (Metric) Diameter (Metric) Cuff
31337508 Large, 9.5 800 mm 0.6 mm 300 mm
31337400 Medium, 8 800 mm 0.4 mm 180 mm
31337436 Medium, 8 800 mm 0.6 mm 180 mm
31337484 Medium, 8 800 mm 0.4 mm 200 mm
31337398 Medium, 8 800 mm 0.6 mm 200 mm
31337454 Medium, 8 800 mm 0.4 mm 225 mm
31337483 Medium, 8 800 mm 0.6 mm 225 mm
31337482 Large, 9.5 800 mm 0.4 mm 160 mm
31337497 Large, 9.5 800 mm 0.6 mm 160 mm
31337429 Large, 9.5 800 mm 0.4 mm 180 mm
31337439 Large, 9.5 800 mm 0.6 mm 200 mm
31337416 Large, 9.5 800 mm 0.4 mm 225 mm
31337440 Large, 9.5 800 mm 0.6 mm 225 mm
31337517 Large, 9.5 800 mm 0.4 mm 250 mm
31337495 Large, 9.5 800 mm 0.6 mm 250 mm
31337424 Large, 9.5 800 mm 0.4 mm 300 mm
31337461 Large, 9.5 920 mm 0.4 mm 200 mm
31337476 Large, 9.5 920 mm 0.6 mm 200 mm
31337494 Large, 9.5 920 mm 0.4 mm 250 mm
31337469 Large, 9.5 920 mm 0.6 mm 250 mm
31337473 X-Large, 11 800 mm 0.4 mm 180 mm
31337396 X-Large, 11 800 mm 0.6 mm 200 mm
31337407 X-Large, 11 800 mm 0.4 mm 225 mm
31337486 X-Large, 11 800 mm 0.6 mm 225 mm
31337493 X-Large, 11 800 mm 0.4 mm 250 mm
31337419 X-Large, 11 800 mm 0.6 mm 250 mm
31337401 X-Large, 11 800 mm 0.4 mm 300 mm
31337457 X-Large, 11 920 mm 0.4 mm 300 mm
31337518 X-Large, 11 800 mm 0.6 mm 300 mm
31337399 X-Large, 11 920 mm 0.4 mm 225 mm
31337467 X-Large, 11 920 mm 0.6 mm 225 mm
31337447 X-Large, 11 920 mm 0.4 mm 250 mm
31337488 X-Large, 11 920 mm 0.6 mm 250 mm
31337462 X-Large, 11 920 mm 0.6 mm 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
34 options
Product Code. 31337508 Supplier AL Glove Systems Supplier No. 1230068101
Add to basket
Add to basket
Flexible, chemically resistant, and impermeable to gas.
AL Glove Systems GSB gloves are made of bromo-butyl rubber (BBR) with high gas impermeability and high resistance to a wide range of polar hydrocarbons such as acids, esters, ketones, and amine derivatives. They are highly flexible with a high sensitivity. The GSB Glove is classified as PAH-free.
  • Impermeable to gas
  • High resistance to the permeation of mainly oxygen, nitrogen, inert gases, etc. during production under inert gas atmosphere in inert boxes for the production of electronic components and microcomponents
Flexible:
  • Offers good sensitivity in handling
  • Resistant to chemicals
  • High resistance to a wide range of chemicals and toxins
PERMEATION EN ISO 374-1:2016/A1:2018
  • A Methanol Protection Level 6
  • B Acetone Protection Level 6
  • I Ethyl acetate Protection Level 3
  • K Sodium hydroxide 40% Protection Level 6
  • L Sulphuric acid 96% Protection Level 6
  • N Acetic acid 99% Protection Level 6
  • O Ammonium hydroxide 25% Protection Level 6
  • T Formaldehyde 37% Protection Level 6
Specifications
Certifications/Compliance (Europe) PPE Category III Directive 2016/425 EC Type
Chlorinated No
Color Black
Hand Ambidextrous
Latex Free Yes
Material Butyl
Product Type Dry Box, Isolator Gloves
Size Large, 9.5
Temperature (English) Max. 194°F
Temperature (Metric) Max 90°C
Thickness (English) 24 mil
Thickness (Metric) 0.6 mm
For Use With (Application) Handling Sensitive or Hazardous Materials in an Inert or Contained Atmosphere
For Use With (Equipment) Dry Box, Isolator, RABS
Grip Design Smooth
Manufacturing/QMS Audit Standards ISO 9001:2015
Packaging Type Carton
Temperature (English) Min -40°F
Temperature (Metric) Min. -40°C
Length (English) 32 in.
Length (Metric) 800 mm
Diameter (English) Cuff 12 In.
Diameter (Metric) Cuff 300 mm