Learn More
Microspec™ Grip Seal Bags
Product Code. 16685482
Change view
Click to view available options
Dimensions (L x W):
2 x 9 in.
3 x 7.5 in.
6 x 9 in.
7.5 x 7.5 in.
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
16685482 7.5 x 7.5 in. Pack of 1000
16655482 2 x 9 in. Pack of 1000
16665482 3 x 7.5 in. Pack of 1000
16675482 6 x 9 in. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16685482 Supplier Microspec™ Supplier No. W8
  • ONLINE EXCLUSIVE PRICE
    87.62 EUR
    75.39 EUR / Pack of 1000
    Save 12.23 EUR
    13% Off
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Pack of 1000 Grip Seal Bags
  • Grip Seal Bag
  • Sterile
  • Pack of 1000
Specifications
Autoclavable Not Autoclavable
Disposable Single-use
For Use With (Application) Sample Transport
Sterility Sterile
Film Description LDPE
No. per Case 1000
No. per Pack 100
Sterilization Method EtO
Dimensions (L x W) 7.5 x 7.5 in.