Learn More
MedChemExpress GRF (1-29) amide (rat)
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Purity | 98.22% |
| CAS | 91826-20-9 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 3473.08 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 16.67 mg/mL (4.80 mM∣Need ultrasonic) |
| Synonym | rGHRH(1-29)NH2 |