Learn More
Greiner Bio-One Blood Collection Tubes
Product Code. 15412550
Change view
Click to view available options
Capacity (Metric):
9 mL
Closure Color:
White/Black
Additive:
None
Tube Size:
16 x 100 mm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Additive Tube Size Unit Size
15412550 9 mL White/Black None 16 x 100 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15412550 Supplier Greiner Bio-One Supplier No. 455001
  • ONLINE EXCLUSIVE PRICE
    14.18 EUR
    11.10 EUR / Pack of 50
    Save 3.08 EUR
    21% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Can be used for CSF determinations
  • No additive tubes
  • Polyethylene Therephthalate
  • Come in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Product Type Evacuated Blood Collection Tube
Additive None
Closure Type Non-ridged Pull Cap
Capacity (Metric) 9 mL
Closure Color White/Black
Tube Size 16 x 100 mm
For Use With (Application) Blood collection