Learn More
Cole-Parmer™ Thermal Transfer Compound, Tube
Product Code. 11840500 Shop All Cole Parmer Products
Change view
Click to view available options
Material:
Grease
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11840500 Grease Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11840500 Supplier Cole-Parmer™ Supplier No. 9378561
Estimated Shipment: 15-02-2027
Thermal Transfer Compound increases thermal conductivity in thermo wells for faster response time.
  • Designed to transfer heat away from heat-generating components.
  • Silicone-free heat sink compound increases thermal conductivity in thermo wells for faster response time in comparison to air.
Specifications
For Use With (Application) Increases thermal conductivity.
Material Grease
Weight (English) 4 oz.
Weight (Metric) 113.4 g
Product Type Thermal Transfer Compound
For Use With (Equipment) Thermo Wells