Learn More
Graphite rod, 2.54cm (1.0in) dia x 61cm (24in) long, 99% (metals basis), Thermo Scientific Chemicals
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Chemical Name or Material | Graphite rod |
| Name Note | 2.54cm (1.0 in.) dia. x 61cm (24 in.) long |
| Quantity | 1 Pc. |
| Weight | ≈590g/pc |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD00144065 |
| Percent Purity | 99% |