Learn More
Thermo Scientific Chemicals Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Click to view available options
Capacity (Metric):
7.5 cc
7.88 cc
Quantity:
36 Pieces
50 Pieces
Unit Size:
Pack of 36
Pack of 50
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Description | Unpurified extruded graphite |
| Material | Graphite |
| For Use With (Application) | Metallurgy |
| Quantity | 50 Pieces |
| Capacity (Metric) | 7.5 cc |
| Product Type | Graphite fusion crucible |
| Shape | Drillpoint |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur