missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Gramicidin

Product Code. 16494987
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16494987 Supplier MedChemExpress Supplier No. HYP0163
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gramicidin is an antimicrobial peptide assembling as channels in membranes and increasing their permeability towards cations.

Specifications

CAS 1405-97-6
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) COVID-19-immunoregulation
Molecular Weight (g/mol) 1882.3 g/mol
Product Type Natural Products
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : 100 mg/mL (53.13 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
Product Title
Select an issue

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.