Learn More
- Home
- Products
- Pipet Products
- Transfer Pipets
- Globe Scientific Graduated Transfer Pipets
Globe Scientific Graduated Transfer Pipets
Product Code. 31923007 Shop All Globe Scientific Products
Change view
Click to view available options
Bulb Draw (Metric):
0.9 mL
1.8 mL
2 mL
2.2 mL
3.2 mL
3.4 mL
3.5 mL
5 mL
Length (Metric):
115 mm
140 mm
145 mm
148 mm
150 mm
155 mm
215 mm
Packaging Type:
10/Pk., 10 Pk/Bag, 4 Bags/Cs.
20/Pk., 20 Pk/Cs.
20/Pk., 25 Pk/Cs.
250/Pk., 10 Pk/Cs.
500/Pk., 10 Pk/Cs.
Individually Wrapped, 100/Pk., 4 Pk/Cs.
Individually Wrapped, 100/Pk., 5 Pk/Cs.
Unit Size:
Pack of 2500
Pack of 400
Pack of 500
Pack of 5000
24 product options available for selection
| Product Code. | Bulb Draw (Metric) | Length (Metric) | Packaging Type | Unit Size |
|---|---|---|---|---|
| 31923007 | 3.2 mL | 155 mm | Individually Wrapped, 100/Pk., 4 Pk/Cs. | Pack of 400 |
| 31922852 | 3.5 mL | 148 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31923684 | 3.5 mL | 148 mm | Individually Wrapped, 100/Pk., 4 Pk/Cs. | Pack of 400 |
| 31922810 | 3.2 mL | 155 mm | 10/Pk., 10 Pk/Bag, 4 Bags/Cs. | Pack of 400 |
| 31923662 | 3.2 mL | 155 mm | 20/Pk., 20 Pk/Cs. | Pack of 400 |
| 31924079 | 0.9 mL | 115 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31923622 | 0.9 mL | 115 mm | Individually Wrapped, 100/Pk., 5 Pk/Cs. | Pack of 500 |
| 31923912 | 0.9 mL | 115 mm | 20/Pk., 25 Pk/Cs. | Pack of 500 |
| 31922995 | 3.4 mL | 150 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31924097 | 3.4 mL | 150 mm | 20/Pk., 20 Pk/Cs. | Pack of 400 |
| 31924070 | 3.4 mL | 150 mm | Individually Wrapped, 100/Pk., 4 Pk/Cs. | Pack of 400 |
| 31924086 | 3.4 mL | 145 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31923564 | 2.2 mL | 140 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31923145 | 2.2 mL | 140 mm | 20/Pk., 20 Pk/Cs. | Pack of 400 |
| 31923737 | 3.4 mL | 145 mm | Individually Wrapped, 100/Pk., 4 Pk/Cs. | Pack of 400 |
| 31922366 | 1.8 mL | 155 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31923874 | 1.8 mL | 155 mm | Individually Wrapped, 100/Pk., 5 Pk/Cs. | Pack of 500 |
| 31921455 | 1.8 mL | 155 mm | 20/Pk., 25 Pk/Cs. | Pack of 500 |
| 31922991 | 2 mL | 155 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31923632 | 2.2 mL | 140 mm | Individually Wrapped, 100/Pk., 4 Pk/Cs. | Pack of 400 |
| 31923009 | 3.4 mL | 145 mm | 10/Pk., 10 Pk/Bag, 4 Bags/Cs. | Pack of 400 |
| 31924074 | 3.4 mL | 145 mm | 20/Pk., 20 Pk/Cs. | Pack of 400 |
| 31924093 | 1.8 mL | 150 mm | 500/Pk., 10 Pk/Cs. | Pack of 5000 |
| 31923008 | 5 mL | 215 mm | 250/Pk., 10 Pk/Cs. | Pack of 2500 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
Denna artikel kan inte returneras.
Se returpolicy
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- Material: Low Density Polyethylene (LDPE)
- Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean.
Specifikationer
Specifikationer
| Autoclavable | Not Autoclavable |
| Bulb Draw (Metric) | 3.2 mL |
| Calibrated | No |
| Material | LDPE |
| Sterility | Sterile |
| Drops/mL | 20 |
| Graduated | Yes |
| Graduations | 3 mL |
| Length (English) | 6.3 in. |
| Packaging Type | Individually Wrapped, 100/Pk., 4 Pk/Cs. |
| Color | Natural |
| Volume (Metric) | 7 mL |
| Length (Metric) | 155 mm |
| Type | Graduated Transfer Pipet |
| Disposable | Single-use |
| Visa mer Visa mindre |
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?