Learn More
Gilac™ Graduated Round Bucket with Stainless Steel Handle
Product Code. 17938931
Change view
Click to view available options
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
17938931 Each
17908661 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17938931 Supplier Gilac™ Supplier No. G615521
Only null left
Add to basket
Add to basket
Graduated Round Bucket with Stainless Steel Handle is a highly durable food bucket with graduation.
  • Food bucket with graduation.
  • Highly durable.
  • Anti-wear skirt for increased product durability.
  • Made from low density polyethylene making it highly resistant.
  • Stainless steel handle.
  • External dimensions include handles.
Specifications
Height (English) 12.6 in.
Height (Metric) 320 mm
Includes Bucket and Handle
Quantity 14 L