Learn More
Hirschmann™ Graduated Pipettes with Piston
Product Code. 10482081
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
Length (Metric):
335 mm
345 mm
360 mm
385 mm
430 mm
475 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Unit Size
10482081 1 mL 335 mm Each
10083900 25 mL 475 mm Each
10073900 10 mL 385 mm Each
10432661 5 mL 360 mm Each
10014740 20 mL 430 mm Each
10698221 2 mL 345 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10482081 Supplier Hirschmann™ Supplier No. 1190133
  • ONLINE EXCLUSIVE PRICE
    28.00 EUR
    24.55 EUR / Pack of 1
    Save 3.45 EUR
    12% Off
Estimated Shipment: 22-12-2026
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Graduated Pipettes with Piston come with amber stain graduation.
  • Graduated to tip
  • Available in a range of capacities
Specifications
Graduated Yes
Graduation Color Amber
Graduation Type Amber graduations permanently with silver diffusion color
Quantity 1 Each
Volume (Metric) 1 mL
Length (Metric) 335 mm
Type Graduated Pipet