Learn More
Fisherbrand™ Grade 605 Very Absorbent Joseph Paper
Product Code. 11700565 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
120 x 120 mm
150 x 150 mm
Format:
Sheet
Packaging Quantity:
200 Pk.
500 Pk.
Unit Size:
Pack of 200
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Format Packaging Quantity Unit Size
11700565 120 x 120 mm Sheet 500 Pk. Pack of 500
11728722 150 x 150 mm Sheet 200 Pk. Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11700565
  • ONLINE EXCLUSIVE PRICE
    20.65 EUR
    19.63 EUR / Pack of 500
    Save 1.02 EUR
    4% Off
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
Used in laboratories and hospitals. Fisherbrand™ Grade 605 Very Absorbent Joseph Paper is thin and not fluffy.
Applications:
  • Cleaning and drying of, glass containers, tubes, flasks, microscopic plates, bottles
Specifications
Length (Metric) 120 mm
Width (Metric) 120 mm
Dimensions (L x W) 120 x 120 mm
Format Sheet
For Use With (Application) Cleaning and drying of, Glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals
Packaging Quantity 500 Pk.
Grade 605