Learn More
MedChemExpress GPRP acetate
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Specifications
Specifications
| Product Type | Biochemical Assay Reagents |
| CAS | 157009-81-9 |
| Synonym | Gly-Pro-Arg-Pro acetate ∣ Pefa 6003 acetate |
| Solubility | H2O : 125 mg/mL (257.45 mM; Need ultrasonic) |
| Content And Storage | Sealed storage, away from moisture ∣ Powder -80°C 2 years, -20°C 1 year ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Shipping Condition | Room temperature |
| Quantity | 5 mg |
| Molecular Weight (g/mol) | 485.53 g/mol |
| Target | NO Synthase ∣ MMP ∣ Endogenous Metabolite |