missing translation for 'onlineSavingsMsg'
Learn More

GPR37 Antibody (436923), Biotin, Novus Biologicals™

Product Code. 18695593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18695593 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18695593 Supplier Novus Biologicals Supplier No. FAB4450B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR37 Monoclonal antibody specifically detects GPR37 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR37
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 436923
Conjugate Biotin
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias EDNRBL, Endothelin B receptor-like protein 1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR37, Ala27-Cys613, Accession # O15354
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2861
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.