missing translation for 'onlineSavingsMsg'
Learn More

GPR32 Antibody - BSA Free, Novus Biologicals™

Product Code. 18718642 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18718642 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18718642 Supplier Novus Biologicals Supplier No. NB10058979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR32 Polyclonal antibody specifically detects GPR32 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR32
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS
Gene Alias G protein-coupled receptor 32, probable G-protein coupled receptor 32
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR32.
Purification Method Immunogen affinity purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2854
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.