missing translation for 'onlineSavingsMsg'
Learn More

GPR171 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Product Code. 18389306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389306 100 μL 100µL
18321046 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18389306 Supplier Novus Biologicals Supplier No. NBP303890100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR171 Polyclonal specifically detects GPR171 in Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR171
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias F730001G15Rik, G protein-coupled receptor 171, G-protein coupled receptor H963, H963platelet activating receptor homolog, probable G-protein coupled receptor 171
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human GPR171 (NP_037440.3).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline GPCR, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 29909
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.