missing translation for 'onlineSavingsMsg'
Learn More

GPCR GPR89 Antibody - BSA Free, Novus Biologicals™

Product Code. 18785560 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18785560 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18785560 Supplier Novus Biologicals Supplier No. NB10058981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPCR GPR89 Polyclonal antibody specifically detects GPCR GPR89 in Human, Mouse, Rat, Porcine, Bovine, Equine, Hamster, Primate, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPCR GPR89
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS
Gene Accession No. P0CG08
Gene Alias FLJ10492, FLJ11696, FLJ36909, FLJ79270, G protein-coupled receptor 89A, Golgi pH regulator A, GPHRA, GPR89, GPR89B, MGC688, MGC87324, protein GPR89, Protein GPR89A, Putative MAPK-activating protein PM01, Putative NF-kappa-B-activating protein 90, SH120, UNQ192
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from cytoplasmic loop of human GPCR GPR89.
Purification Method Immunogen affinity purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653519
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.