Learn More
Invitrogen™ GPCR/G-protein complex-stabilizing scFv Recombinant Mouse Monoclonal Antibody (scFv16)
Product Code. 17194679
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17194679 1 mg 1mg
17184679 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17194679 Supplier Invitrogen™ Supplier No. 7039761MG
Mouse Recombinant Monoclonal Antibody
The scFv antibody stabilizes GPCR/G-protein complexes for cryo-EM of active structures. It recognizes an interface between G alpha and G beta/gamma subunits, and confers resistance to nucleotide exchange-triggered dissociation of the heterotrimeric G-protein. In addition to members of the Gi/o family, this antibody fragment can also be applied to chimeric variants and stabilize other G-protein subtypes including Gs complexes.
TRUSTED_SUSTAINABILITY
Specifications
Antigen GPCR/G-protein complex-stabilizing scFv
Applications Not Available
Classification Recombinant Monoclonal
Clone scFv16
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation 25mM HEPES with 10% glycerol and no preservative; pH 7.5
Host Species Mouse
Immunogen Rhodopsin/Gi1.
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid