missing translation for 'onlineSavingsMsg'
Learn More

Gossypol, MP Biomedicals™

Product Code. 11451122 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451122 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11451122 Supplier MP Biomedicals Supplier No. 0219521083

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 303-45-7
Molecular Formula C30H30O8
Molecular Weight (g/mol) 518.56
MDL Number MFCD00017352
InChI Key QBKSWRVVCFFDOT-UHFFFAOYSA-N
Synonym gossypol, --gossypol, +-gossypol, pogosin, tash 1, racemic-gossypol, r---gossypol, r-gossypol, +--gossypol, +/--gossypol
PubChem CID 3503
ChEBI CHEBI:28584
IUPAC Name 1,1',6,6',7,7'-hexahydroxy-3,3'-dimethyl-5,5'-bis(propan-2-yl)-[2,2'-binaphthalene]-8,8'-dicarbaldehyde
SMILES CC(C)C1=C(O)C(O)=C(C=O)C2=C(O)C(=C(C)C=C12)C1=C(O)C2=C(C=O)C(O)=C(O)C(C(C)C)=C2C=C1C

Specifications

Color Yellow
Quantity 250 mg
Formula Weight 518.6
Physical Form Solid
Chemical Name or Material Gossypol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.