Learn More
Gosselin™ Polystrene Petri Dish
BIOPHARMA
Product Code. 15258043
Change view
Click to view available options
Diameter (Metric):
55 mm
140 mm
Height (Metric):
14.2 mm
15 mm
20.6 mm
Vent:
0
3
6
Sterility:
Aseptic
Sterile
Unit Size:
Pack of 1620
Pack of 176
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) Vent Sterility Unit Size
15258043 140 mm 20.6 mm 3 Sterile Pack of 176
11758573 55 mm 14.2 mm 6 Aseptic Pack of 1620
15268043 140 mm 20.6 mm 3 Aseptic Pack of 176
11718593 - 15 mm 0 Sterile Pack of 176
11788573 55 mm 14.2 mm 0 Sterile Pack of 1620
11778573 55 mm 14.2 mm 0 Aseptic Pack of 1620
11708593 140 mm 20.6 mm 0 Aseptic Pack of 176
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15258043 Lieferant Gosselin Lieferanten-Nr. BP14306
  • ONLINE EXCLUSIVE PRICE
    139.00 EUR
    113.30 EUR / Pack of 176
    Save 25.70 EUR
    18% Off
Estimated Shipment: 08-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Crystal polystyrene dishes, with optimal flatness.
  • Appropriate for aseptic production
  • Sterilized by irradiation
  • Flat bottom
TRUSTED_SUSTAINABILITY
Spezifikation
Certifications/Compliance EC N°1935/2004
Closure Included Closure Included
Color Natural
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
Well Count 1
For Use With (Application) For use in clinical, environment, industries, microbiology
Vent 3
Height (Metric) 20.6 mm
Diameter (Metric) 140 mm
Mehr anzeigen Weniger anzeigen